4-Azidocinnamaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 3-(4-Azidophenyl)-2-propenal, 3-(4-Azidophenyl)acrylaldehyde

OR937942-10G OR937942-1G OR937942-5G
APOSOR937942-10GEA 0 EUR
APOSOR937942-10G APOSOR937942-1G APOSOR937942-5G
4-Azidocinnamaldehyde 95%
4-Azidocinnamaldehyde
Formel: C₉H₇N₃O
Molecular Weight: 173.18 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 22736-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR