N-Carbobenzoxyhydroxylamine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Hydroxycarbamic acid Benzyl Ester, N-Cbz-hydroxylamine, Benzyl N-Hydroxycarbamate, Benzylhydroxycarbamate

OR937984-1G OR937984-25G OR937984-5G OR937984-10G
APOSOR937984-1GEA 21.9 EUR
APOSOR937984-1G APOSOR937984-25G APOSOR937984-5G APOSOR937984-10G
N-Carbobenzoxyhydroxylamine 98%
N-Carbobenzoxyhydroxylamine
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
Schmelzpunkt: 65…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010642
CAS-Nummer: 3426-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR