Valeryl bromide 95%

Lieferant: Apollo Scientific

Synonyme: Valeryl Bromide, Valeroyl Bromide, Pentanoyl bromide

OR937986-5G
APOSOR937986-5GEA 151 EUR
APOSOR937986-5G
Valeryl bromide 95%
Valeryl bromide
Formel: C₅H₉BrO
Molecular Weight: 165.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059467
CAS-Nummer: 1889-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR