Butyl crotonate 95%
Lieferant: Apollo Scientific
Synonyme:
Crotonic Acid Butyl Ester
OR937997-100G
OR937997-25G
OR937997-500G
APOSOR937997-100GEA
270
EUR
APOSOR937997-100G
APOSOR937997-25G
APOSOR937997-500G
Butyl crotonate 95%
Butyl crotonate
Formel:
C₈H₁₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053794 CAS-Nummer: 7299-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...