(R)-(+)-1-Ethyl-2-Pyrrolidincarboxylsäure-Ethylester 95%

Lieferant: Apollo Scientific

Synonyme: (R)-(+)-1-Ethyl-2-pyrrolidinecarboxylic Acid Ethyl Ester, (+)-1-Ethyl-D-proline Ethyl Ester, Et-D-Pro-OEt

OR938006-1G OR938006-5G
APOSOR938006-1GEA 203 EUR
APOSOR938006-1G APOSOR938006-5G
(R)-(+)-1-Ethyl-2-Pyrrolidincarboxylsäure-Ethylester 95%
Ethyl (R)-(+)-1-ethyl-2-pyrrolidinecarboxylate
Formel: C₉H₁₇NO₂
Molecular Weight: 171.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00671533
CAS-Nummer: 381670-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR