3-Aminoimidazo[1,2-a]pyridin 95%

Lieferant: Apollo Scientific

OR938028-1G OR938028-250MG
APOSOR938028-1GEA 464 EUR
APOSOR938028-1G APOSOR938028-250MG
3-Aminoimidazo[1,2-a]pyridin 95%
3-Aminoimidazo[1,2-a]pyridin
Formel: C₇H₇N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06660421
CAS-Nummer: 28036-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR