2-Amino-2-oxazoline hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
4,5-Dihydrooxazol-2-amine hydrochloride
OR938063-1G
OR938063-5G
APOSOR938063-1GEA
255
EUR
APOSOR938063-1G
APOSOR938063-5G
2-Amino-2-oxazoline hydrochloride 95%
2-Amino-2-oxazoline hydrochloride
Formel:
C₃H₆N₂O Molecular Weight: 122,55 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060178 CAS-Nummer: 375855-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...