Homogentisäure gamma-Lacton 95%
Lieferant: Apollo Scientific
Synonyme:
2,3-Dihydro-5-hydroxybenzofuran-2-one, 2,5-Dihydroxyphenylacetic Acid γ-Lactone, 5-Hydroxybenzofuran-2(3H)-one, 5-Hydroxy-2-coumaranone
OR938120-5G
APOSOR938120-5GEA
130
EUR
APOSOR938120-5G
Homogentisäure gamma-Lacton 95%
Homogentisic acid-gamma-lactone
Formel:
C₈H₆O₃ Molecular Weight: 150,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005857 CAS-Nummer: 2688-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...