3-Chlor-2,2-dimethylpropansäure 97%
Lieferant: Apollo Scientific
Synonyme:
3-Chloro-2,2-dimethylpropionic Acid, 3-Chloro-2,2-dimethylpropanoic acid
OR938167-25G
APOSOR938167-25GEA
177
EUR
APOSOR938167-25G
3-Chlor-2,2-dimethylpropansäure 97%
3-Chlor-2,2-dimethylpropansäure
Formel:
C₅H₉ClO₂ Molecular Weight: 136,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004195 CAS-Nummer: 13511-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...