3-(Allyloxy)propansäure 95%
Lieferant: Apollo Scientific
OR938182-10G
OR938182-1G
OR938182-5G
APOSOR938182-10GEA
580
EUR
APOSOR938182-10G
APOSOR938182-1G
APOSOR938182-5G
3-(Allyloxy)propansäure 95%
3-(Allyloxy)propansäure
Formel:
C₆H₁₀O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039537 CAS-Nummer: 22577-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...