(R)-(+)-Citronellal 90%

Lieferant: Apollo Scientific

Synonyme: (R)-(+)-3,7-Dimethyl-6-octenal

OR938258-1G OR938258-5G
APOSOR938258-1GEA 145 EUR
APOSOR938258-1G APOSOR938258-5G
(R)-(+)-Citronellal 90%
(R)-(+)-Citronellal
Formel: C₁₀H₁₈O
Molecular Weight: 154,25 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2385-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR