5-(Isopropylamino)-1-pentanol 97%

Lieferant: Apollo Scientific

Synonyme: 5-(Isopropylamino)pentan-1-ol

OR938297-1G OR938297-5G
APOSOR938297-1GEA 105 EUR
APOSOR938297-1G APOSOR938297-5G
5-(Isopropylamino)-1-pentanol 97%
5-(Isopropylamino)-1-pentanol
Formel: C₈H₁₉NO
Molecular Weight: 145.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14708174
CAS-Nummer: 40447-21-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR