2,4-Dimethoxy-3-methylbenzaldehyd 95%

Lieferant: Apollo Scientific

OR938299-5G
APOSOR938299-5GEA 201 EUR
APOSOR938299-5G
2,4-Dimethoxy-3-methylbenzaldehyd 95%
2,4-Dimethoxy-3-methylbenzaldehyde
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2005 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011489
CAS-Nummer: 7149-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR