2,6-Naphthalindiyldimethanol 95%
Lieferant: Apollo Scientific
Synonyme:
Naphthalene-2,6-diyldimethanol
OR938513-1G
OR938513-5G
APOSOR938513-1GEA
206
EUR
APOSOR938513-1G
APOSOR938513-5G
2,6-Naphthalindiyldimethanol 95%
2,6-Bis(hydroxymethyl)naphthalene
Formel:
C₁₂H₁₂O₂ Molecular Weight: 188.23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11870109 CAS-Nummer: 5859-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...