2-Methylbenzhydryl chloride 98%

Lieferant: Apollo Scientific

Synonyme: 2-(alpha-Chlorobenzyl)toluene, Chloro(2-methylphenyl)phenylmethane

OR938518-5G
APOSOR938518-5GEA 59.6 EUR
APOSOR938518-5G
2-Methylbenzhydryl chloride 98%
2-Methylbenzhydryl chloride
Formel: C₁₄H₁₃Cl
Molecular Weight: 216,71 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00972765
CAS-Nummer: 41870-52-4
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR