Ethyl-2,4,6-trimethylbenzoat 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl Mesitylenecarboxylate, Ethyl 2,4,6-trimethylbenzoate, Mesitylenecarboxylic Acid Ethyl Ester, 2,4,6-Trimethylbenzoic Acid Ethyl Ester

OR938538-25G
APOSOR938538-25GEA 140 EUR
APOSOR938538-25G
Ethyl-2,4,6-trimethylbenzoat 95%
Ethyl-2,4,6-trimethylbenzoate
Formel: C₁₂H₁₆O₂
Molecular Weight: 192,26 g/mol
Siedepunkt: 121 °C/6,75mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00015439
CAS-Nummer: 1754-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR