2,4-Dimethoxy-1,3,5-triazin 95%
Lieferant: Apollo Scientific
OR938544-1G
OR938544-5G
APOSOR938544-1GEA
42.2
EUR
APOSOR938544-1G
APOSOR938544-5G
2,4-Dimethoxy-1,3,5-triazin 95%
2,4-Dimethoxy-1,3,5-triazine
Formel:
C₅H₇N₃O₂ Molecular Weight: 141,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20257908 CAS-Nummer: 1898-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...