Methyl-3-methoxypropionat 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-methoxypropionate

OR938562-100G OR938562-1KG OR938562-500G
APOSOR938562-100GEA 125 EUR
APOSOR938562-100G APOSOR938562-1KG APOSOR938562-500G
Methyl-3-methoxypropionat 95%
Methyl-3-methoxypropionat
Formel: C₅H₁₀O₃
Molecular Weight: 118,1311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059085
CAS-Nummer: 3852-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR