3-(4-Chlorophenyl)-2-oxopropanoic acid 97%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-a-oxo-benzenepropanoic acid

OR938565-1G OR938565-250MG
APOSOR938565-1GEA 654 EUR
APOSOR938565-1G APOSOR938565-250MG
3-(4-Chlorophenyl)-2-oxopropanoic acid 97%
3-(4-Chlorophenyl)-2-oxopropanoic acid
Formel: C₉H₇ClO₃
Molecular Weight: 198,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00969474
CAS-Nummer: 3617-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR