(±)-3,7-Dimethyl-3-octanol 95%
Lieferant: Apollo Scientific
Synonyme:
(±)-Tetrahydrolinalool
OR938576-100G
OR938576-500G
APOSOR938576-100GEA
23.2
EUR
APOSOR938576-100G
APOSOR938576-500G
(±)-3,7-Dimethyl-3-octanol 95%
(±)-3,7-Dimethyl-3-octanol
Formel:
C₁₀H₂₂O Molecular Weight: 158,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004482 CAS-Nummer: 78-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...