3-Bromphenethylbromid 97%

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-3-(2-bromoethyl)benzene

OR938612-1G OR938612-5G
APOSOR938612-1GEA 52.2 EUR
APOSOR938612-1G APOSOR938612-5G
3-Bromphenethylbromid 97%
3-Bromphenethylbromid
Formel: C₈H₈Br₂
Molecular Weight: 263,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784359
CAS-Nummer: 40422-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR