Methyl (R)-(+)-3-bromoisobutyrate 95%

Lieferant: Apollo Scientific

Synonyme: (R)-Methyl 3-bromo-2-methylpropanoate, Methyl (R)-(+)-3-Bromo-2-methylpropionate, (R)-(+)-3-Bromoisobutyric Acid Methyl Ester, (R)-(+)-3-Bromo-2-Methylpropionic Acid Methyl Ester

OR938686-1G OR938686-5G
APOSOR938686-1GEA 214 EUR
APOSOR938686-1G APOSOR938686-5G
Methyl (R)-(+)-3-bromoisobutyrate 95%
Methyl (R)-(+)-3-bromoisobutyrate
Formel: C₅H₉BrO₂
Molecular Weight: 181,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010639
CAS-Nummer: 110556-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR