4-(2-Hydroxyethyl)cyclohexanol (cis/trans-Gemisch) 95%
Lieferant: Apollo Scientific
Synonyme: 2-(4-Hydroxycyclohexyl)ethanol, 4-Hydroxycyclohexaneethanol
OR938745-1G OR938745-250MG OR938745-5G
APOSOR938745-1GEA 306 EUR
APOSOR938745-1G APOSOR938745-250MG APOSOR938745-5G
4-(2-Hydroxyethyl)cyclohexanol (cis/trans-Gemisch) 95%
4-(2-Hydroxyethyl)cyclohexanol (cis- and trans- mixture)
Formel: C₈H₁₆O₂
Molecular Weight: 144.21 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 74058-21-2