3,3-Dimethylbutylamine 95%
Lieferant: Apollo Scientific
OR938756-1G
OR938756-5G
APOSOR938756-1GEA
179
EUR
APOSOR938756-1G
APOSOR938756-5G
3,3-Dimethylbutylamine 95%
3,3-Dimethylbutylamine
Formel:
C₆H₁₅N Lagertemperatur: Raumtemperatur |
CAS-Nummer:
15673-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...