2-(Methylthio)benzaldehyd 90%
Lieferant: Apollo Scientific
|
Achtung
|
OR938789-1G
OR938789-5G
APOSOR938789-1GEA
56.7
EUR
APOSOR938789-1G
APOSOR938789-5G
2-(Methylthio)benzaldehyd 90%
2-(Methylthio)benzaldehyde
Formel:
C₈H₈OS Molecular Weight: 152,2135 g/mol Siedepunkt: 141…143 °C Dichte: 1,18 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00196822 CAS-Nummer: 7022-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...