N-Methyl-N-1-naphthylamine 95%

Lieferant: Apollo Scientific

Synonyme: N-Methylnaphthalen-1-amine

OR938798-5G
APOSOR938798-5GEA 115 EUR
APOSOR938798-5G
N-Methyl-N-1-naphthylamine 95%
N-Methyl-N-1-naphthylamine
Formel: C₁₁H₁₁N
Molecular Weight: 157,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00972770
CAS-Nummer: 2216-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR