1-Phenylethylacetat 95%
Lieferant: Apollo Scientific
Synonyme:
Styrallylacetat, Essigsäure(1-phenylethyl)ester, α-Methylbenzylacetat
OR938810-500G
APOSOR938810-500GEA
78.3
EUR
APOSOR938810-500G
1-Phenylethylacetat 95%
1-Phenylethylacetat
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008858 CAS-Nummer: 93-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...