1,1-Ethandiyl-diacetat 95%

Lieferant: Apollo Scientific

Synonyme: 1,1-Diacetoxyethane, Ethylidene Diacetate

OR938851-1G OR938851-25G OR938851-5G OR938851-100G
APOSOR938851-1GEA 0 EUR
APOSOR938851-1G APOSOR938851-25G APOSOR938851-5G APOSOR938851-100G
1,1-Ethandiyl-diacetat 95%
1,1-Ethanediol diacetate
Formel: C₆H₁₀O₄
Molecular Weight: 146,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014980
CAS-Nummer: 542-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR