trans-4-(Hydroxymethyl)cyclohexanol 95%

Lieferant: Apollo Scientific

OR938871-1G OR938871-250MG
APOSOR938871-1GEA 104 EUR
APOSOR938871-1G APOSOR938871-250MG
trans-4-(Hydroxymethyl)cyclohexanol 95%
trans-4-(Hydroxymethyl)cyclohexanol
Formel: C₇H₁₄O₂
Molecular Weight: 130.1900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11036248
CAS-Nummer: 3685-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR