1-Phenyl-1H-pyrazol-3-amine 95%

Lieferant: Apollo Scientific

OR938962-100MG OR938962-250MG
APOSOR938962-100MGEA 23.2 EUR
APOSOR938962-100MG APOSOR938962-250MG
1-Phenyl-1H-pyrazol-3-amine 95%
1-Phenyl-1H-pyrazol-3-amine
Formel: C₉H₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00100520
CAS-Nummer: 1128-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR