2-Methyl-1-phenyl-2-butanol 95%

Lieferant: Apollo Scientific

OR939054-25G OR939054-5G
APOSOR939054-25GEA 377 EUR
APOSOR939054-25G APOSOR939054-5G
2-Methyl-1-phenyl-2-butanol 95%
2-Methyl-1-phenyl-2-butanol
Formel: C₁₁H₁₆O
Molecular Weight: 164,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01708473
CAS-Nummer: 772-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR