2-Methyl-5-(-1-pyrrolidinylsulphonyl)benzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Methyl-5-(pyrrolidin-1-ylsulphonyl)phenyl)boronic acid

OR9391-1G OR9391-500MG OR9391-250MG
APOSOR9391-1GEA 277 EUR
APOSOR9391-1G APOSOR9391-500MG APOSOR9391-250MG
2-Methyl-5-(-1-pyrrolidinylsulphonyl)benzeneboronic acid 98%
2-Methyl-5-(-1-pyrrolidinylsulphonyl)benzeneboronic acid
Formel: C₁₁H₁₆BNO₄S
Molecular Weight: 269.12 g/mol
Schmelzpunkt: 126…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08056365
CAS-Nummer: 871333-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR