(1R,5S)-8-Azabicyclo[3.2.1]octan-3-one

Lieferant: Apollo Scientific
Achtung

OR939255-1G OR939255-5G
APOSOR939255-1GEA 264 EUR
APOSOR939255-1G APOSOR939255-5G
(1R,5S)-8-Azabicyclo[3.2.1]octan-3-one
(1R,5S)-8-Azabicyclo[3.2.1]octan-3-one
Formel: C₇H₁₁NO
Molecular Weight: 125.17 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD14279165
CAS-Nummer: 5632-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR