Furan-2-acetic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-(Furan-2-yl)acetic acid

OR939287-1G
APOSOR939287-1GEA 102 EUR
APOSOR939287-1G
Furan-2-acetic acid 95%
Furan-2-acetic acid
Formel: C₆H₆O₃
Molecular Weight: 126,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798036
CAS-Nummer: 2745-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR