Iodomesitylen Diacetat 98%

Lieferant: Apollo Scientific

Synonyme: 2,4,6-Trimethyl(diacetoxyiodo)benzene, 2,4,6-Trimethyliodobenzene Diacetate, 2-(Diacetoxyiodo)mesitylene

OR939426-1G OR939426-25G OR939426-5G
APOSOR939426-1GEA 0 EUR
APOSOR939426-1G APOSOR939426-25G APOSOR939426-5G
Iodomesitylen Diacetat 98%
Iodomesitylene diacetate
Formel: C₁₃H₁₇IO₄
Molecular Weight: 364,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00674196
CAS-Nummer: 33035-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR