4-Ethynylbenzyl alcohol 99%
Lieferant: Apollo Scientific
Synonyme:
(4-Ethynylphenyl)methanol
OR939454-10G
OR939454-1G
OR939454-5G
APOSOR939454-10GEA
207
EUR
APOSOR939454-10G
APOSOR939454-1G
APOSOR939454-5G
4-Ethynylbenzyl alcohol 99%
4-Ethynylbenzyl alcohol
Formel:
C₉H₈O Molecular Weight: 132.16 g/mol Schmelzpunkt: 40…44 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05664205 CAS-Nummer: 10602-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...