N-Formylsaccharin

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Oxobenzo[d]isothiazole-2(3H)-carbaldehyde 1,1-dioxide

OR939711-1G OR939711-25G OR939711-5G
APOSOR939711-1GEA 50.9 EUR
APOSOR939711-1G APOSOR939711-25G APOSOR939711-5G
N-Formylsaccharin
N-Formylsaccharin
Formel: C₈H₅NO₄S
Molecular Weight: 211,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23380208
CAS-Nummer: 50978-45-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR