5-Phenyl-1,2,4-oxadiazol-3-amine 95%
Lieferant: Apollo Scientific
OR939721-1G
OR939721-250MG
OR939721-500MG
APOSOR939721-1GEA
696
EUR
APOSOR939721-1G
APOSOR939721-250MG
APOSOR939721-500MG
5-Phenyl-1,2,4-oxadiazol-3-amine 95%
5-Phenyl-1,2,4-oxadiazol-3-amine
Formel:
C₈H₇N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00197235 CAS-Nummer: 7788-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...