4-Methoxyphenethylbromid 97%

Lieferant: Apollo Scientific

Synonyme: 4-(2-Bromethyl)anisol, 1-Brom-2-(4-methoxyphenyl)ethan

OR939726-5G
APOSOR939726-5GEA 66.7 EUR
APOSOR939726-5G
4-Methoxyphenethylbromid 97%
4-Methoxyphenethylbromid
Formel: C₉H₁₁BrO
Molecular Weight: 215,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07369786
CAS-Nummer: 14425-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR