8-Aminoquinoline-N-oxide 95%
Lieferant: Apollo Scientific
OR939750-1G
OR939750-250MG
APOSOR939750-1GEA
319
EUR
APOSOR939750-1G
APOSOR939750-250MG
8-Aminoquinoline-N-oxide 95%
8-Aminoquinoline-N-oxide
Formel:
C₉H₈N₂O Molecular Weight: 160,18 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
92339-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...