3,3,5-Trimethylcyclohexanon 95%

Lieferant: Apollo Scientific

OR939767-100G OR939767-10G OR939767-25G
APOSOR939767-100GEA 574 EUR
APOSOR939767-100G APOSOR939767-10G APOSOR939767-25G
3,3,5-Trimethylcyclohexanon 95%
3,3,5-Trimethylcyclohexanon
Formel: C₉H₁₆O
Molecular Weight: 140.23 g/mol
Siedepunkt: 188,8
Schmelzpunkt: -10
Lagertemperatur: Raumtemperatur
MDL: MFCD00019466
CAS-Nummer: 873-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR