4-(Phenoxymethyl)benzoesäure 97%

Lieferant: Apollo Scientific

OR939781-1G OR939781-250MG
APOSOR939781-1GEA 169 EUR
APOSOR939781-1G APOSOR939781-250MG
4-(Phenoxymethyl)benzoesäure 97%
4-(Phenoxymethyl)benzoic acid
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.2433 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00623268
CAS-Nummer: 31719-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR