N-Benzyl-N,N'-dimethylethylenediamine 96%

Lieferant: Apollo Scientific

OR939818-1G OR939818-25G OR939818-5G
APOSOR939818-1GEA 132 EUR
APOSOR939818-1G APOSOR939818-25G APOSOR939818-5G
N-Benzyl-N,N'-dimethylethylenediamine 96%
N-Benzyl-N,N'-dimethylethylenediamine
Formel: C₁₁H₁₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00014856
CAS-Nummer: 102-11-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR