2-Isopropenyl-5-methyl-5-vinyltetrahydrofuran (mixture of isomers) 95%

Lieferant: Apollo Scientific

Synonyme: Dehydroxylinalool Oxide

OR939864-25G OR939864-5G
APOSOR939864-25GEA 255 EUR
APOSOR939864-25G APOSOR939864-5G
2-Isopropenyl-5-methyl-5-vinyltetrahydrofuran (mixture of isomers) 95%
2-Isopropenyl-5-methyl-5-vinyltetrahydrofuran (mixture of isomers)
Formel: C₁₀H₁₆O
Molecular Weight: 152,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00065422
CAS-Nummer: 13679-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR