5-Brom-2-methoxybenzolsulfonamid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR9399-1G
OR9399-250MG
OR9399-5G
APOSOR9399-1GEA
140
EUR
APOSOR9399-1G
APOSOR9399-250MG
APOSOR9399-5G
5-Brom-2-methoxybenzolsulfonamid 95%
5-Brom-2-methoxybenzolsulfonamid
Formel:
C₇H₈BrNO₃S Schmelzpunkt: 151…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06808831 CAS-Nummer: 23095-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...