N-Allyl-N-tert-butylamine 95%

Lieferant: Apollo Scientific

Synonyme: N-tert-Butylallylamine

OR939901-25G OR939901-5G
APOSOR939901-25GEA 214 EUR
APOSOR939901-25G APOSOR939901-5G
N-Allyl-N-tert-butylamine 95%
N-Allyl-N-tert-butylamine
Formel: C₇H₁₅N
Molecular Weight: 113,20 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 16486-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR