Tetrahydrofuran-2-Essigsäure Ethylester 95%

Lieferant: Apollo Scientific

Synonyme: Tetrahydrofuran-2-acetic Acid Ethyl Ester

OR939909-10G OR939909-1G OR939909-5G
APOSOR939909-10GEA 806 EUR
APOSOR939909-10G APOSOR939909-1G APOSOR939909-5G
Tetrahydrofuran-2-Essigsäure Ethylester 95%
Ethyl tetrahydrofuran-2-acetate
Formel: C₈H₁₄O₃
Molecular Weight: 158.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059735
CAS-Nummer: 2434-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR