5-Brom-N-butyl-2-methoxybenzolsulfonamid 98%

Lieferant: Apollo Scientific

OR9402-1G OR9402-5G
APOSOR9402-1GEA 139 EUR
APOSOR9402-1G APOSOR9402-5G
5-Brom-N-butyl-2-methoxybenzolsulfonamid 98%
5-Brom-N-butyl-2-methoxybenzolsulfonamid
Formel: C₁₁H₁₆BrNO₃S
Schmelzpunkt: 93…96 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08056377
CAS-Nummer: 871269-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR