5-Brom-N,N-dimethyl-2-methoxybenzolsulfonamid 97%

Lieferant: Apollo Scientific

OR9405-1G OR9405-5G
APOSOR9405-1GEA 139 EUR
APOSOR9405-1G APOSOR9405-5G
5-Brom-N,N-dimethyl-2-methoxybenzolsulfonamid 97%
5-Brom-N,N-dimethyl-2-methoxybenzolsulfonamid
Formel: C₉H₁₂BrNO₃S
Schmelzpunkt: 163…166 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06808833
CAS-Nummer: 871269-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR