5-Brom-N-ethyl-2-methoxybenzolsulfonamid 95%

Lieferant: Apollo Scientific

OR9406-1G OR9406-5G
APOSOR9406-1GEA 139 EUR
APOSOR9406-1G APOSOR9406-5G
5-Brom-N-ethyl-2-methoxybenzolsulfonamid 95%
5-Brom-N-ethyl-2-methoxybenzolsulfonamid
Formel: C₉H₁₂BrNO₃S
Schmelzpunkt: 121…123 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06003173
CAS-Nummer: 717892-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR